Tisk…

2-Bromo-6-iodo-3-methoxypyridine

Dodavatel: Apollo Scientific

OR61061-10G OR61061-1G OR61061-5G
APOSOR61061-10GEA 19280 CZK
APOSOR61061-10G APOSOR61061-1G APOSOR61061-5G
2-Bromo-6-iodo-3-methoxypyridine
2-Bromo-6-iodo-3-methoxypyridine
Vzorec: C₆H₅BrINO
Bod tání: 99…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909341
CAS číslo: 321535-37-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR