2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
DavePhos, 2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl, 2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine, 2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl
OR61059-25G
OR61059-5G
OR61059-100G
APOSOR61059-25GEA
3480
CZK
APOSOR61059-25G
APOSOR61059-5G
APOSOR61059-100G
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl
Usage subject to US Patents 6307087 and 6395916. This product is intended for research and development purposes only.
Vzorec:
C₂₆H₃₆NP MW: 393,55 g/mol Bod tání: 121…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD02183572 CAS číslo: 213697-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...