Tisk…

Diethoxy(dimethyl)silane

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Dimethyldiethoxysilane

OR61048-100G OR61048-2.5KG OR61048-500G
APOSOR61048-100GEA 622 CZK
APOSOR61048-100G APOSOR61048-2.5KG APOSOR61048-500G
Diethoxy(dimethyl)silane
Diethoxy(dimethyl)silane
Vzorec: C₆H₁₆O₂Si
Bod varu: 111…113 °C
Bod tání: –70 °C
Hustota: 0,865 g/cm³ (25 °C)
Bod vzplanutí: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009068
CAS číslo: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR