3-(Carboxymethyl)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(3-Boronophenyl)acetic acid
OR6104-250MG
OR6104-1G
APOSOR6104-250MGEA
3770
CZK
APOSOR6104-250MG
APOSOR6104-1G
3-(Carboxymethyl)phenylboronic acid 98%
3-(Carboxymethyl)phenylboronic acid
Vzorec:
C₈H₉BO₄ MW: 179,9700 g/mol Bod tání: 188…190 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08704669 CAS číslo: 914397-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...