Tisk…

3-Chloro-5-formylpyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloro-pyridine-3-carbaldehyde, 5-Chloronicotinaldehyde

OR61039-1G OR61039-250MG OR61039-5G
APOSOR61039-1GEA 1580 CZK
APOSOR61039-1G APOSOR61039-250MG APOSOR61039-5G
3-Chloro-5-formylpyridine
3-Chloro-5-formylpyridine
Vzorec: C₆H₄ClNO
MW: 141,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457773
CAS číslo: 113118-82-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR