4-Methyl-3-pyridineboronic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Methylpyridin-3-yl)boronic acid, 4-Methylpyridine-3-boronic acid
OR6103-1G
OR6103-5G
OR6103-25G
APOSOR6103-1GEA
1080
CZK
APOSOR6103-1G
APOSOR6103-5G
APOSOR6103-25G
4-Methyl-3-pyridineboronic acid 96%
4-Methyl-3-pyridineboronic acid
Vzorec:
C₆H₈BNO₂ MW: 136.94 g/mol Bod tání: 102…106 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788244 CAS číslo: 148546-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...