Tisk…

5-Bromo-2-cyano-4-methylpyridine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-4-methylpicolinonitrile, 5-Bromo-4-methylpyridine-2-carbonitrile

OR61014-5G
APOSOR61014-5GEA 3540 CZK
APOSOR61014-5G
5-Bromo-2-cyano-4-methylpyridine 97%
5-Bromo-2-cyano-4-methylpyridine
Vzorec: C₇H₅BrN₂
MW: 197,0300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375090
CAS číslo: 886364-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR