Trimethyl orthobutyrate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Trimethylorthobutyrate, 1,1,1-Trimethoxybutane, Orthobutyric Acid Trimethyl Ester
OR61007-5ML
OR61007-25ML
APOSOR61007-5MLEA
2370
CZK
APOSOR61007-5ML
APOSOR61007-25ML
Trimethyl orthobutyrate
Trimethyl orthobutyrate
Vzorec:
C₇H₁₆O₃ MW: 148.20 g/mol Bod varu: 146 °C Hustota: 0.93 g/cm³ (25 °C) Bod vzplanutí: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008480 CAS číslo: 43083-12-1 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...