6-Isoquinolinol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Hydroxyisoquinoline, Isoquinolin-6-ol
OR6099-1G
OR6099-250MG
OR6099-500MG
APOSOR6099-1GEA
1450
CZK
APOSOR6099-1G
APOSOR6099-250MG
APOSOR6099-500MG
6-Isoquinolinol 98%
6-Isoquinolinol
Vzorec:
C₉H₇NO MW: 145,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04114860 CAS číslo: 7651-82-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...