Tetrahydrothiopyran-4-carbonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR6086-1G
OR6086-5G
APOSOR6086-1GEA
1990
CZK
APOSOR6086-1G
APOSOR6086-5G
Tetrahydrothiopyran-4-carbonitrile 97%
Tetrahydrothiopyran-4-carbonitrile
Vzorec:
C₆H₉NS Storage Temperature: Ambient |
MDL Number:
MFCD09025909 CAS číslo: 195503-40-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...