2-(4-Benzoylphenyl)-2-phenylacetonitrile 95%
Dodavatel: Apollo Scientific
OR6064-1G
OR6064-500MG
APOSOR6064-1GEA
9200
CZK
APOSOR6064-1G
APOSOR6064-500MG
2-(4-Benzoylphenyl)-2-phenylacetonitrile 95%
2-(4-Benzoylphenyl)-2-phenylacetonitrile
Vzorec:
C₂₁H₁₅NO Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00231979 CAS číslo: 338791-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...