Tisk…

(1-Amino-2-chloroethylidene)malononitrile 95%

Dodavatel: Apollo Scientific

OR6053-10G OR6053-1G OR6053-5G
APOSOR6053-10GEA 20170 CZK
APOSOR6053-10G APOSOR6053-1G APOSOR6053-5G
(1-Amino-2-chloroethylidene)malononitrile 95%
(1-Amino-2-chloroethylidene)malononitrile
Vzorec: C₅H₄ClN₃
Bod tání: 157 °C
Storage Temperature: Ambient
CAS číslo: 118645-77-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR