Tetrahydroxydiborane 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR60243-25G
OR60243-5G
OR60243-100G
APOSOR60243-25GEA
2320
CZK
APOSOR60243-25G
APOSOR60243-5G
APOSOR60243-100G
Tetrahydroxydiborane 97%
Tetrahydroxydiborane
Vzorec:
H₄B₂O₄ Bod tání: 143…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663888 CAS číslo: 13675-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...