Tisk…

2-[4-(Bromomethyl)phenyl]propionic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR60242-100G OR60242-25G
APOSOR60242-100GEA 12630 CZK
APOSOR60242-100G APOSOR60242-25G
2-[4-(Bromomethyl)phenyl]propionic acid 97%
2-[4-(Bromomethyl)phenyl]propionic acid
Vzorec: C₁₀H₁₁BrO₂
Bod tání: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD02093445
CAS číslo: 111128-12-2
UN: 1759
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR