4-Bromo-2-chloro-6-nitroaniline
Dodavatel: Apollo Scientific
|
Varování
|
OR60213-1G
OR60213-250MG
OR60213-5G
APOSOR60213-1GEA
910
CZK
APOSOR60213-1G
APOSOR60213-250MG
APOSOR60213-5G
4-Bromo-2-chloro-6-nitroaniline
4-Bromo-2-chloro-6-nitroaniline
Vzorec:
C₆H₄BrClN₂O₂ MW: 251,4652 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03426655 CAS číslo: 34033-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...