3-Amino-2-cyano-5-methylpyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR60181-1G
OR60181-5G
APOSOR60181-1GEA
1370
CZK
APOSOR60181-1G
APOSOR60181-5G
3-Amino-2-cyano-5-methylpyridine
3-Amino-2-cyano-5-methylpyridine
Vzorec:
C₇H₇N₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11227185 CAS číslo: 1001635-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...