Tisk…

1-(Tetrahydro-2H-pyran-4-yl)ethanone 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Acetyltetrahydropyran

OR60176-1G OR60176-25G OR60176-5G
APOSOR60176-1GEA 622 CZK
APOSOR60176-1G APOSOR60176-25G APOSOR60176-5G
1-(Tetrahydro-2H-pyran-4-yl)ethanone 97%
1-(Tetrahydro-2H-pyran-4-yl)ethanone
Vzorec: C₇H₁₂O₂
MW: 128,17 g/mol
Bod varu: 90…94 °C/15mmHg
Hustota: 1,024 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08704647
CAS číslo: 137052-08-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR