Tisk…

4-Ethyl-1H-pyrazol-3-amine

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-4-ethyl-1H-pyrazole

OR60148-1G OR60148-5G
APOSOR60148-1GEA 6190 CZK
APOSOR60148-1G APOSOR60148-5G
4-Ethyl-1H-pyrazol-3-amine
4-Ethyl-1H-pyrazol-3-amine
Vzorec: C₅H₉N₃
MW: 111,148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06797570
CAS číslo: 43024-15-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR