Tisk…

3-Bromo-5-methylpicolinic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-5-methylpyridine-2-carboxylic acid

OR60145-1G OR60145-5G OR60145-10G
APOSOR60145-1GEA 2030 CZK
APOSOR60145-1G APOSOR60145-5G APOSOR60145-10G
3-Bromo-5-methylpicolinic acid 97%
3-Bromo-5-methylpicolinic acid
Vzorec: C₇H₆BrNO₂
MW: 216,0320 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214206
CAS číslo: 1211515-68-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR