Tisk…

6-Bromo-2-chloromethylimidazo[1,2-a]pyridine

Dodavatel: Apollo Scientific

OR60134-1G OR60134-250MG OR60134-500MG OR60134-2.5G OR60134-5G
APOSOR60134-1GEA 14520 CZK
APOSOR60134-1G APOSOR60134-250MG APOSOR60134-500MG APOSOR60134-2.5G APOSOR60134-5G
6-Bromo-2-chloromethylimidazo[1,2-a]pyridine
6-Bromo-2-chloromethylimidazo[1,2-a]pyridine
Vzorec: C₈H₆BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD06739243
CAS číslo: 136117-72-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR