Tisk…

5-Chloro-2-methoxyphenol

Dodavatel: Apollo Scientific
Varování

OR60133-1G OR60133-250MG OR60133-5G
APOSOR60133-1GEA 1910 CZK
APOSOR60133-1G APOSOR60133-250MG APOSOR60133-5G
5-Chloro-2-methoxyphenol
5-Chloro-2-methoxyphenol
Vzorec: C₇H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11100133
CAS číslo: 3743-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR