Tisk…

Methyl-3-iodo-1H-indazole-4-carboxylate

Dodavatel: Apollo Scientific
Varování

OR60119-1G OR60119-100MG
APOSOR60119-1GEA 2490 CZK
APOSOR60119-1G APOSOR60119-100MG
Methyl-3-iodo-1H-indazole-4-carboxylate
Methyl-3-iodo-1H-indazole-4-carboxylate
Vzorec: C₉H₇IN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07781588
CAS číslo: 885521-54-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR