Tisk…

Methyl-3-iodo-1H-indazole-5-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 3-iodo-1H-indazole-5-carboxylate

OR60118-100MG OR60118-1G OR60118-250MG
APOSOR60118-100MGEA 622 CZK
APOSOR60118-100MG APOSOR60118-1G APOSOR60118-250MG
Methyl-3-iodo-1H-indazole-5-carboxylate
Methyl-3-iodo-1H-indazole-5-carboxylate
Vzorec: C₉H₇IN₂O₂
MW: 302.0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739144
CAS číslo: 885271-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR