Tisk…

5-Chloro-3-iodo-1H-indazole

Dodavatel: Apollo Scientific
Varování

OR60117-1G OR60117-500MG
APOSOR60117-1GEA 910 CZK
APOSOR60117-1G APOSOR60117-500MG
5-Chloro-3-iodo-1H-indazole
5-Chloro-3-iodo-1H-indazole
Vzorec: C₇H₄ClIN₂
MW: 278.4800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739146
CAS číslo: 351456-45-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR