1H-Indazole-6-boronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1H-Indazol-6-ylboronic acid, 6-Indazolyboronic acid, 1H-Indazol-6-yl-6-boronic acid
OR60112-1G
OR60112-5G
APOSOR60112-1GEA
1450
CZK
APOSOR60112-1G
APOSOR60112-5G
1H-Indazole-6-boronic acid
1H-Indazole-6-boronic acid
Vzorec:
C₇H₇BN₂O₂ MW: 161,9500 g/mol Bod tání: 180…184 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04972419 CAS číslo: 885068-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...