Tisk…

Cyclobutylboronic acid

Dodavatel: Apollo Scientific
Varování

OR60086-5G OR60086-1G
APOSOR60086-5GEA 6220 CZK
APOSOR60086-5G APOSOR60086-1G
Cyclobutylboronic acid
Cyclobutylboronic acid
Vzorec: C₄H₉BO₂
MW: 99.9241 g/mol
Bod tání: 118…123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798108
CAS číslo: 849052-26-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR