2-Cyano-5-bromomethylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-(Bromomethyl)picolinonitrile, 5-(Bromomethyl)pyridine-2-carbonitrile
OR60075-1G
OR60075-5G
APOSOR60075-1GEA
8040
CZK
APOSOR60075-1G
APOSOR60075-5G
2-Cyano-5-bromomethylpyridine
2-Cyano-5-bromomethylpyridine
Vzorec:
C₇H₅BrN₂ MW: 197,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09037857 CAS číslo: 308846-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...