Tisk…

4-Isoquinolinecarboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Isoquinoline-4-carboxylic acid

OR60073-5G
APOSOR60073-5GEA 5390 CZK
APOSOR60073-5G
4-Isoquinolinecarboxylic acid 98%
4-Isoquinolinecarboxylic acid
Vzorec: C₁₀H₇NO₂
MW: 173,1681 g/mol
Bod tání: 265…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00094344
CAS číslo: 7159-36-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR