Tisk…

2,6-Dichloronicotinonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,6-Dichloropyridine-3-carbonitrile, 2,6-Dichloro-3-cyanopyridine

OR60049-25G
APOSOR60049-25GEA 2940 CZK
APOSOR60049-25G
2,6-Dichloronicotinonitrile 97%
2,6-Dichloronicotinonitrile
Vzorec: C₆H₂Cl₂N₂
MW: 173,00 g/mol
Bod tání: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD03411341
CAS číslo: 40381-90-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR