Pyridazin-4-amine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Aminopyridazine
OR60046-1G
OR60046-5G
APOSOR60046-1GEA
2280
CZK
APOSOR60046-1G
APOSOR60046-5G
Pyridazin-4-amine 97%
Pyridazin-4-amine
Vzorec:
C4H5N3 MW: 95,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00233975 CAS číslo: 20744-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...