Tisk…

2-Chloropyridine-3-sulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR60041-250MG OR60041-1G
APOSOR60041-250MGEA 622 CZK
APOSOR60041-250MG APOSOR60041-1G
2-Chloropyridine-3-sulphonyl chloride 97%
2-Chloropyridine-3-sulphonyl chloride
Vzorec: C₅H₃Cl₂NO₂S
Bod tání: 42…43 °C
Storage Temperature: Ambient
MDL Number: MFCD05664916
CAS číslo: 6684-06-6
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR