(+/-)-Equol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(4-Hydroxyphenyl)chroman-7-ol, (R,S)-Equol, 4′,7-Dihydroxyisoflavane, 7-Hydroxy-3-(4-hydroxyphenyl)chroman, (±)-Equol, 4',7-Isoflavandiol, 3-(4-Hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol
OR60026-1G
OR60026-5G
APOSOR60026-1GEA
744
CZK
APOSOR60026-1G
APOSOR60026-5G
(+/-)-Equol
(+/-)-Equol
Vzorec:
C₁₅H₁₄O₃ MW: 242.27 g/mol Bod tání: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016662 CAS číslo: 94105-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...