2-Chloropyrimidine-4-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR60023-1G
OR60023-250MG
APOSOR60023-1GEA
622
CZK
APOSOR60023-1G
APOSOR60023-250MG
2-Chloropyrimidine-4-carboxylic acid
2-Chloropyrimidine-4-carboxylic acid
Vzorec:
C₅H₃ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06739041 CAS číslo: 149849-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...