Tisk…

4-Bromo-2-methyl-2H-indazole

Dodavatel: Apollo Scientific
Varování

OR60016-1G OR60016-250MG
APOSOR60016-1GEA 2860 CZK
APOSOR60016-1G APOSOR60016-250MG
4-Bromo-2-methyl-2H-indazole
4-Bromo-2-methyl-2H-indazole
Vzorec: C₈H₇BrN₂
MW: 211,0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870046
CAS číslo: 590417-93-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR