4-Bromo-1-methyl-1H-indazole 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR60015-1G
APOSOR60015-1GEA
910
CZK
APOSOR60015-1G
4-Bromo-1-methyl-1H-indazole 98%
4-Bromo-1-methyl-1H-indazole
Vzorec:
C₈H₇BrN₂ MW: 211,0586 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09870042 CAS číslo: 365427-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...