(S)-(+)-sec-Butylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(S)-(+)-2-Aminobutane, (S)-(+)-2-Butylamine
OR60000-250MG
APOSOR60000-250MGEA
6350
CZK
APOSOR60000-250MG
(S)-(+)-sec-Butylamine 98%
(S)-(+)-sec-Butylamine
Vzorec:
C₄H₁₁N MW: 73,14 g/mol Bod varu: 63 °C Bod tání: –105 °C Hustota: 0,722 g/cm³ (25 °C) Bod vzplanutí: –19 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064417 CAS číslo: 513-49-5 EINECS: 208-164-7 UN: 2733 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...