(R)-(-)-sec-Butylamine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(R)-(-)-2-Butylamine, (R)-(-)-2-Aminobutane
OR59999-1G
OR59999-250MG
OR59999-5G
APOSOR59999-1GEA
3940
CZK
APOSOR59999-1G
APOSOR59999-250MG
APOSOR59999-5G
(R)-(-)-sec-Butylamine
(R)-(-)-sec-Butylamine
Vzorec:
C₄H₁₁N Bod varu: 63 °C Hustota: 0,722 g/cm³ (25 °C) Bod vzplanutí: –19 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064416 CAS číslo: 13250-12-9 UN: 2733 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...