Tisk…

5-Methylfuran-2-sulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR59996-1G OR59996-250MG
APOSOR59996-1GEA 21760 CZK
APOSOR59996-1G APOSOR59996-250MG
5-Methylfuran-2-sulphonyl chloride 95%
5-Methylfuran-2-sulphonyl chloride
Vzorec: C₅H₅ClO₃S
Storage Temperature: Ambient
MDL Number: MFCD11109334
CAS číslo: 69815-95-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR