Cyclobutylamine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Cyclobutylamine, Cyclobutanamine, Aminocyclobutane
OR59991-25G
OR59991-5G
OR59991-100G
OR59991-1G
APOSOR59991-25GEA
6620
CZK
APOSOR59991-25G
APOSOR59991-5G
APOSOR59991-100G
APOSOR59991-1G
Cyclobutylamine
Cyclobutylamine
Vzorec:
C₄H₉N MW: 71,12 g/mol Bod varu: 81…83 °C Hustota: 0,833 g/cm³ (25 °C) Bod vzplanutí: –4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001328 CAS číslo: 2516-34-9 EINECS: 219-736-0 UN: 2733 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...