Ethyl-α-bromophenylacetate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Ethyl alpha-bromophenylacetate
OR59981-10G
OR59981-250G
OR59981-50G
APOSOR59981-10GEA
4570
CZK
APOSOR59981-10G
APOSOR59981-250G
APOSOR59981-50G
Ethyl-α-bromophenylacetate
Ethyl-α-bromophenylacetate
Vzorec:
C₁₀H₁₁BrO₂ MW: 243,10 g/mol Bod varu: 164 °C/30mmHg Hustota: 1,389 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013536 CAS číslo: 2882-19-1 EINECS: 220-735-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...