Tetrahydro-2H-pyran-4-carbothioamide 90%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Tetrahydro-2H-pyran-4-thiocarboxamide
OR59976-1G
OR59976-250MG
APOSOR59976-1GEA
9330
CZK
APOSOR59976-1G
APOSOR59976-250MG
Tetrahydro-2H-pyran-4-carbothioamide 90%
Tetrahydro-2H-pyran-4-carbothioamide
Vzorec:
C₆H₁₁NOS MW: 145,22 g/mol Bod tání: 162…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD10700048 CAS číslo: 88571-77-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...