Tisk…

Potassium thioacetate

Dodavatel: Apollo Scientific
Varování

Synonyms: Potassium ethanethioate

OR59966-100G OR59966-25G
APOSOR59966-100GEA 1240 CZK
APOSOR59966-100G APOSOR59966-25G
Potassium thioacetate
Potassium thioacetate
Vzorec: C₂H₃KOS
Storage Temperature: Ambient
MDL Number: MFCD00137704
CAS číslo: 10387-40-3
EINECS: 233-848-7
UN: 3335
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR