N-(Diphenylmethylene)glycine ethyl ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl (diphenylmethylenamino)acetate, Ethyl 2-((diphenylmethylene)amino)acetate, Ethyl-2-((diphenylmethylene)amino)acetate, Ethyl N-(diphenylmethylene)glycinate
OR59938-25G
OR59938-5G
OR59938-100G
APOSOR59938-25GEA
622
CZK
APOSOR59938-25G
APOSOR59938-5G
APOSOR59938-100G
N-(Diphenylmethylene)glycine ethyl ester
N-(Diphenylmethylene)glycine ethyl ester
Vzorec:
C₁₇H₁₇NO₂ MW: 267,32 g/mol Bod tání: 51…53 °C Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010590 CAS číslo: 69555-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...