Tisk…

Methyl pyrimidine-2-carboxylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Pyrimidine-2-carboxylic acid methyll ester

OR59919-5G OR59919-100G OR59919-25G
APOSOR59919-5GEA 622 CZK
APOSOR59919-5G APOSOR59919-100G APOSOR59919-25G
Methyl pyrimidine-2-carboxylate 97%
Methyl pyrimidine-2-carboxylate
Vzorec: C₆H₆N₂O₂
MW: 138,1200 g/mol
Bod tání: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD03426929
CAS číslo: 34253-03-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR