4-(Dimethylcarbamoyl)phenylboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-(N,N-Dimethylaminocarbonyl)phenylboronic acid, pinacol ester, 4-(Dimethylcarbamoyl)phenylboronic Acid Pinacol Ester
OR59906-1G
APOSOR59906-1GEA
10650
CZK
APOSOR59906-1G
4-(Dimethylcarbamoyl)phenylboronic acid pinacol ester
4-(Dimethylcarbamoyl)phenylboronic acid pinacol ester
Vzorec:
C₁₅H₂₂BNO₃ MW: 275,1511 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05863908 CAS číslo: 400727-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...