Diethyl (4-methoxybenzyl)phosphonate
Dodavatel: Apollo Scientific
|
Varování
|
OR59905-10G
OR59905-50G
APOSOR59905-10GEA
3840
CZK
APOSOR59905-10G
APOSOR59905-50G
Diethyl (4-methoxybenzyl)phosphonate
Diethyl (4-methoxybenzyl)phosphonate
Vzorec:
C₁₂H₁₉O₄P Bod varu: 136…138 °C/0,7mmHg Hustota: 1,5 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00015662 CAS číslo: 1145-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...