Tisk…

(3,4,5-Trimethoxyphenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,4,5-Trimethoxyphenylacetic acid

OR5990-100G OR5990-5G
APOSOR5990-100GEA 4300 CZK
APOSOR5990-100G APOSOR5990-5G
(3,4,5-Trimethoxyphenyl)acetic acid 98%
(3,4,5-Trimethoxyphenyl)acetic acid
Vzorec: (C₆H₂(OCH₃)₃)CH₂COOH
MW: 226,23 g/mol
Bod tání: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00004336
CAS číslo: 951-82-6
EINECS: 213-456-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR