Cyclopropylacetic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR59893-25G
OR59893-5G
APOSOR59893-25GEA
6620
CZK
APOSOR59893-25G
APOSOR59893-5G
Cyclopropylacetic acid
Cyclopropylacetic acid
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Bod varu: 189…191 °C Hustota: 1,076 g/cm³ (25 °C) Bod vzplanutí: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041544 CAS číslo: 5239-82-7 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...