Tisk…

(3,4-Dimethoxyphenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Homoveratric acid, 3,4-Dimethoxyphenylacetic acid

OR5989-500G
APOSOR5989-500GEA 2980 CZK
APOSOR5989-500G
(3,4-Dimethoxyphenyl)acetic acid 98%
(3,4-Dimethoxyphenyl)acetic acid
Vzorec: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196,2 g/mol
Bod tání: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004335
CAS číslo: 93-40-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR