(2-Hydroxyphenyl)acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Hydroxyphenylacetic acid
OR5987-25G
OR5987-100G
OR5987-500G
APOSOR5987-25GEA
622
CZK
APOSOR5987-25G
APOSOR5987-100G
APOSOR5987-500G
(2-Hydroxyphenyl)acetic acid 98%
(2-Hydroxyphenyl)acetic acid
Vzorec:
(C₆H₄OH)CH₂COOH Bod tání: 145…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004323 CAS číslo: 614-75-5 EINECS: 210-393-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...